Skip to Content
Merck
Anti-PI4KB antibody produced in rabbit Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-PI4KB antibody produced in rabbit

Anti-PI4KB antibody produced in rabbit Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-NPIK, Anti-PI4Kβ, Anti-PI4K-β, Anti-PI4K92, Anti-PIK4CB, Anti-Phosphatidylinositol 4-kinase β, Anti-PtdIns 4-kinase β

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique