Anti-Mouse IgG (Fab specific)–Alkaline Phosphatase antibody produced in goat
Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Recombinant Host
Antibody Form
Research Area
Conjugate
Post Translational Modification
Non-reactive Species
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing