Skip to Content
Merck
Antimony(III) telluride powder, −325 mesh, 99.96% trace metals basis

Antimony(III) telluride

Antimony(III) telluride powder, −325 mesh, 99.96% trace metals basis
Synonym(s):
Antimony sesquitelluride, Antimony telluride, Diantimony tritelluride
Linear Formula:
Sb2Te3
CAS Number:
Molecular Weight:
626.32

Products

Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing