Skip to Content
Merck
Anti-Human IgG (γ-chain specific)−Peroxidase antibody produced in goat IgG fraction of antiserum, buffered aqueous solution

Anti-Human IgG (γ-chain specific)−Peroxidase antibody produced in goat

Anti-Human IgG (γ-chain specific)−Peroxidase antibody produced in goat IgG fraction of antiserum, buffered aqueous solution

Products

Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Application
Post Translational Modification
Species Reactivity
Technique