Skip to Content
Merck
Anti-Human IgG (Fc specific)−Biotin antibody, Mouse monoclonal clone HP-6017, purified from hybridoma cell culture

Anti-Human IgG (Fc specific)−Biotin antibody, Mouse monoclonal

Anti-Human IgG (Fc specific)−Biotin antibody, Mouse monoclonal clone HP-6017, purified from hybridoma cell culture
Synonym(s):
Monoclonal Anti-Human IgG (Fc specific)

Products

Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing