![Anti-fd Bacteriophage−Biotin antibody produced in rabbit ~3.5 mg/mL, IgG fraction of antiserum](/deepweb/assets/sigmaaldrich/product/images/221/319/aad00b13-70d7-4517-9bb0-03d44dbd56e2/640/aad00b13-70d7-4517-9bb0-03d44dbd56e2.jpg)
Anti-fd Bacteriophage−Biotin antibody produced in rabbit
![Anti-fd Bacteriophage−Biotin antibody produced in rabbit ~3.5 mg/mL, IgG fraction of antiserum](/deepweb/assets/sigmaaldrich/product/images/221/319/aad00b13-70d7-4517-9bb0-03d44dbd56e2/640/aad00b13-70d7-4517-9bb0-03d44dbd56e2.jpg)
Synonym(s):
Anti-fd Bacteriophage
Products
Product Category
Brand
Biological Source
Clonality
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing