![Anti-EGFR antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/402/556/2cab16d4-7e72-4cef-ac83-0ddbca58a32a/640/2cab16d4-7e72-4cef-ac83-0ddbca58a32a.jpg)
Anti-EGFR antibody produced in rabbit
![Anti-EGFR antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/402/556/2cab16d4-7e72-4cef-ac83-0ddbca58a32a/640/2cab16d4-7e72-4cef-ac83-0ddbca58a32a.jpg)
Synonym(s):
Anti-Epidermal growth factor receptor precursor antibody produced in rabbit, Anti-Receptor tyrosine-protein kinase ErbB-1 antibody produced in rabbit, ERBB1, Epidermal growth factor receptor, Epidermal growth factor receptor precursor, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Application
Post Translational Modification
Species Reactivity
Technique
Showing 1-13 of 13
Compare
Product Number
Description
Pricing