![Anti-ACSL4 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/453/119/65573111-dd1c-4ded-a54b-452a4338c4c9/640/65573111-dd1c-4ded-a54b-452a4338c4c9.jpg)
Anti-ACSL4 antibody produced in rabbit
![Anti-ACSL4 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/453/119/65573111-dd1c-4ded-a54b-452a4338c4c9/640/65573111-dd1c-4ded-a54b-452a4338c4c9.jpg)
Synonym(s):
Anti-ACS4, Anti-Acyl-CoA synthetase long-chain family member 4, Anti-FACL4, Anti-LACS4, Anti-MRX63, Anti-MRX68, Anti-LACS 4 antibody produced in rabbit, Anti-Long-chain acyl-CoA synthetase 4 antibody produced in rabbit, Anti-Long-chain-acyl-CoA synthetase 4 antibody produced in rabbit, Anti-Long-chain-fatty-acid–CoA ligase 4 antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing