Skip to Content
Merck
Anthraquinone phyproof® Reference Substance

Anthraquinone

Anthraquinone phyproof® Reference Substance
Synonym(s):
Anthraquinone
Empirical Formula (Hill Notation):
C14H8O2
CAS Number:
Molecular Weight:
208.21

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing