Skip to Content
Merck
Amyloid β-Protein Fragment 25-35 ≥97% (HPLC)

Amyloid β-Protein Fragment 25-35

Amyloid β-Protein Fragment 25-35 ≥97% (HPLC)
Empirical Formula (Hill Notation):
C45H81N13O14S
CAS Number:
Molecular Weight:
1060.27

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing