![Amitriptyline metabolite, (±)-E-10-hydroxylated- analytical standard](/deepweb/assets/sigmaaldrich/product/structures/971/218/b03d2d05-bbe8-4a52-8044-90dbc7a20c7f/640/b03d2d05-bbe8-4a52-8044-90dbc7a20c7f.png)
Amitriptyline metabolite, (±)-E-10-hydroxylated-
![Amitriptyline metabolite, (±)-E-10-hydroxylated- analytical standard](/deepweb/assets/sigmaaldrich/product/structures/971/218/b03d2d05-bbe8-4a52-8044-90dbc7a20c7f/640/b03d2d05-bbe8-4a52-8044-90dbc7a20c7f.png)
Synonym(s):
(±)-(E)-5-[3-(Dimethylamino)propylidene]-10,11-dihydro-5H-dibenzo[a,d]cyclohepten-10-ol
Empirical Formula (Hill Notation):
C20H23NO
CAS Number:
Molecular Weight:
293.40
Products
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing