![Amikacin impurity I European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/334/142/6634a947-5a50-4822-bd7b-c53e7e157216/640/6634a947-5a50-4822-bd7b-c53e7e157216.png)
Amikacin impurity I
![Amikacin impurity I European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/334/142/6634a947-5a50-4822-bd7b-c53e7e157216/640/6634a947-5a50-4822-bd7b-c53e7e157216.png)
Synonym(s):
(S)-(−)-4-Amino-2-hydroxybutyric acid
Linear Formula:
H2NCH2CH2CH(OH)CO2H
CAS Number:
Molecular Weight:
119.12
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing