Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum-magnesium alloy, Al97.5Mg2.5
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
GF12844964
Aluminum-magnesium alloy, Al97.5Mg2.5,
2m wire, 0.4mm diameter, as drawn, 97.5%
Expand
View Pricing
GF34897911
Aluminum-magnesium alloy, Al97.5Mg2.5,
5m wire, 0.5mm diameter, annealed, 97.5%
Expand
View Pricing
GF00527240
Aluminum-magnesium alloy, Al97.5Mg2.5,
2m wire, 0.5mm diameter, annealed, 97.5%
Expand
View Pricing
GF32560309
Aluminum-magnesium alloy, Al97.5Mg2.5,
10m wire, 0.4mm diameter, as drawn, 97.5%
Expand
View Pricing
GF53250072
Aluminum-magnesium alloy, Al97.5Mg2.5,
1m wire, 0.4mm diameter, as drawn, 97.5%
Expand
View Pricing
GF94688353
Aluminum-magnesium alloy, Al97.5Mg2.5,
20m wire, 0.4mm diameter, as drawn, 97.5%
Expand
View Pricing
GF35328900
Aluminum-magnesium alloy, Al97.5Mg2.5,
10m wire, 0.5mm diameter, annealed, 97.5%
Expand
View Pricing
GF13595766
Aluminum-magnesium alloy, Al97.5Mg2.5,
1m wire, 0.5mm diameter, annealed, 97.5%
Expand
View Pricing
GF26603119
Aluminum-magnesium alloy, Al97.5Mg2.5,
5m wire, 0.4mm diameter, as drawn, 97.5%
Expand
View Pricing
GF06602824
Aluminum-magnesium alloy, Al97.5Mg2.5,
20m wire, 0.5mm diameter, annealed, 97.5%
Expand
View Pricing