Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminium
Synonym(s):
Aluminium
Empirical Formula (Hill Notation):
Al
CAS Number:
7429-90-5
Molecular Weight:
26.98
Products
Brand
Sigma-Aldrich (2)
Supelco (2)
Formula Weight
Physical Form
Product Line
Quality Segment
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
1.01056
Aluminium,
fine powder, stabilized about 2% fat
Expand
View Pricing
1.01057
Aluminium,
(foil) for analysis 0.3 mm thickness, 30 mm width EMSURE
®
Expand
View Pricing
8.20051
Aluminium,
(granulated) for synthesis
Expand
View Pricing
8.14917
Aluminium,
grit
Expand
View Pricing