Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluchrom O - Resistance Alloy, Fe70Cr25Al5
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-20 of 20
Compare
Product Number
Description
Pricing
GF41665259
Aluchrom O - Resistance Alloy, Fe70Cr25Al5,
250m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF39537041
Aluchrom O - Resistance Alloy, Fe70Cr25Al5,
2m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF19547758
Aluchrom O - Resistance Alloy, Fe70Cr25Al5,
1000m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF95577351
Aluchrom O - Resistance Alloy, Fe70Cr25Al5,
100m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF51500517
Aluchrom O - Resistance Alloy, Fe70Cr25Al5,
10m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF96245736
Aluchrom O - Resistance Alloy, Fe70Cr25Al5,
500m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF94224090
Aluchrom O - Resistance Alloy, Fe70Cr25Al5,
5000m wire, 0.125mm diameter, annealed
Expand
View Pricing
GF61553796
Aluchrom O - Resistance Alloy, Fe70Cr25Al5,
2000m wire, 0.125mm diameter, annealed
Expand
View Pricing
GF82092438
Aluchrom O - Resistance Alloy, Fe70Cr25Al5,
1000m wire, 0.125mm diameter, annealed
Expand
View Pricing
GF15056737
Aluchrom O - Resistance Alloy, Fe70Cr25Al5,
250m wire, 0.125mm diameter, annealed
Expand
View Pricing
GF34221372
Aluchrom O - Resistance Alloy, Fe70Cr25Al5,
25m wire, 0.125mm diameter, annealed
Expand
View Pricing
GF88333320
Aluchrom O - Resistance Alloy, Fe70Cr25Al5,
50m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF66306663
Aluchrom O - Resistance Alloy, Fe70Cr25Al5,
10m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF42130459
Aluchrom O - Resistance Alloy, Fe70Cr25Al5,
5m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF30716298
Aluchrom O - Resistance Alloy, Fe70Cr25Al5,
1000m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF08920458
Aluchrom O - Resistance Alloy, Fe70Cr25Al5,
100m wire, 0.125mm diameter, annealed
Expand
View Pricing
GF08156381
Aluchrom O - Resistance Alloy, Fe70Cr25Al5,
500m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF37564612
Aluchrom O - Resistance Alloy, Fe70Cr25Al5,
50m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF78012008
Aluchrom O - Resistance Alloy, Fe70Cr25Al5,
5m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF77671367
Aluchrom O - Resistance Alloy, Fe70Cr25Al5,
50m wire, 0.125mm diameter, annealed
Expand
View Pricing