![Aloin A phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/220/420/5a1104a5-38e8-42cd-9cd2-4582ee4acf90/640/5a1104a5-38e8-42cd-9cd2-4582ee4acf90.png)
Aloin A
![Aloin A phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/220/420/5a1104a5-38e8-42cd-9cd2-4582ee4acf90/640/5a1104a5-38e8-42cd-9cd2-4582ee4acf90.png)
Synonym(s):
Aloin, 1,8-Dihydroxy-10-(β-D-glucopyranosyl)-3-(hydroxymethyl)-9(10H)-anthracenone, 10-β-D-Glucopyranosyl-1,8-dihydroxy-3-(hydroxymethyl)-9(10H)-anthracenone, Aloin A, Barbaloin
Empirical Formula (Hill Notation):
C21H22O9
CAS Number:
Molecular Weight:
418.39
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing