Skip to Content
Merck
Allyl isothiocyanate PESTANAL®, analytical standard

Allyl isothiocyanate

Allyl isothiocyanate PESTANAL®, analytical standard
Synonym(s):
AITC, Oil of mustard
Linear Formula:
CH2=CHCH2NCS
CAS Number:
Molecular Weight:
99.15
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing