Skip to Content
Merck
Aldicarb-(N-methyl-13C,d3, carbamoyl-13C) sulfone ≥98 atom %, ≥98% (CP)

Aldicarb-(N-methyl-13C,d3, carbamoyl-13C) sulfone

Aldicarb-(N-methyl-13C,d3, carbamoyl-13C) sulfone ≥98 atom %, ≥98% (CP)
Empirical Formula (Hill Notation):
13C2C5D3H11N2O4S
CAS Number:
Molecular Weight:
227.27

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing