![Advantame® analytical standard](/deepweb/assets/sigmaaldrich/product/structures/416/618/56903b62-596d-4ac9-88db-29bbfe81e0cd/640/56903b62-596d-4ac9-88db-29bbfe81e0cd.png)
Advantame
![Advantame® analytical standard](/deepweb/assets/sigmaaldrich/product/structures/416/618/56903b62-596d-4ac9-88db-29bbfe81e0cd/640/56903b62-596d-4ac9-88db-29bbfe81e0cd.png)
Synonym(s):
Advantame®, N-[N-[3-(3-Hydroxy-4-methoxyphenyl)propyl]-α-L-aspartyl]-L-phenylalanine 1-methyl ester monohydrate
Empirical Formula (Hill Notation):
C24H30N2O7 · H2O
CAS Number:
Molecular Weight:
476.52
Products
Brand
Formula Weight
Manufacturer
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing