Skip to Content
Merck
Aceclofenac for peak identification European Pharmacopoeia (EP) Reference Standard

Aceclofenac for peak identification

Aceclofenac for peak identification European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Aceclofenac, 2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid carboxymethyl ester;
Empirical Formula (Hill Notation):
C16H13Cl2NO4
CAS Number:
Molecular Weight:
354.18

Products

Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1