Skip to Content
AT
EN
Back
9-Fluorenone
Synonym(s):
9-Fluorenone
Empirical Formula (Hill Notation):
C
13
H
8
O
CAS Number:
486-25-9
Molecular Weight:
180.20
Products
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
F1506
9-Fluorenone,
98%
Expand
View Pricing
8.03977
9-Fluorenone,
for synthesis
Expand
View Pricing