
6-tert-Butyl-2,4-dimethylphenol

Empirical Formula (Hill Notation):
C12H18O
CAS Number:
Molecular Weight:
178.27
Products
Brand
Formula Weight
Manufacturer
Product Line
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing