Skip to Content
Merck
6-Methoxy-2-benzoxazolinone 97%

6-Methoxy-2-benzoxazolinone

6-Methoxy-2-benzoxazolinone 97%
Empirical Formula (Hill Notation):
C8H7NO3
CAS Number:
Molecular Weight:
165.15

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing