5-Bromo-2-nitropyridine
Empirical Formula (Hill Notation):
C5H3BrN2O2
CAS Number:
Molecular Weight:
202.99
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing