Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
5-Aminovaleric acid
Synonym(s):
5-Aminopentanoic acid, Homopiperidinic acid
Linear Formula:
NH
2
(CH
2
)
4
CO
2
H
CAS Number:
660-88-8
Molecular Weight:
117.15
EC Number:
211-544-5
Products
Brand
Sigma-Aldrich (1)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Amino Acid Type
Reaction Type
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
123188
5-Aminovaleric acid,
97%, for peptide synthesis
Expand
View Pricing