Skip to Content
Merck
5-Aminotetrazole monohydrate 97%

5-Aminotetrazole monohydrate

5-Aminotetrazole monohydrate 97%
Synonym(s):
Tetrazol-5-amine
Empirical Formula (Hill Notation):
CH3N5 · H2O
CAS Number:
Molecular Weight:
103.08

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing