Skip to Content
Merck
vibrant-m

5-Amino-2,4,6-triiodo-N-methylisophthalamic acid

vibrant-m
Synonym(s):
3-Amino-2,4,6-triiodo-5-(methylcarbamoyl)benzoic acid
Empirical Formula (Hill Notation):
C9H7I3N2O3
CAS Number:
Molecular Weight:
571.88

Products

Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1