
5-Amino-2,4,6-triiodoisophthalic acid

Linear Formula:
H2NC6I3-1,3-(CO2H)2
Products
Formula Weight
Melting Point (°C)
Purity
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing