Skip to Content
Merck
4-(Trifluoromethyl)phenylboronic acid ≥95.0%

4-(Trifluoromethyl)phenylboronic acid

4-(Trifluoromethyl)phenylboronic acid ≥95.0%
Synonym(s):
α,α,α-Trifluoro-p-tolylboronic acid, 4-(Trifluoromethyl)benzeneboronic acid, [p-(Trifluoromethyl)phenyl]boronic acid
Linear Formula:
CF3C6H4B(OH)2
CAS Number:
Molecular Weight:
189.93

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing