Skip to Content
Merck
4-(Trifluoromethyl)benzenesulfonyl chloride 97%

4-(Trifluoromethyl)benzenesulfonyl chloride

4-(Trifluoromethyl)benzenesulfonyl chloride 97%
Linear Formula:
CF3C6H4SO2Cl
CAS Number:
Molecular Weight:
244.62

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing