![4-Methyl-N-propyl-3-[[(2RS)-2-(propylamino)propanoyl]amino]thiophene-2-carboxamide hydrochloride certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/277/493/7651cc5d-47a5-4017-836e-5a3c52875481/640/7651cc5d-47a5-4017-836e-5a3c52875481.png)
4-Methyl-N-propyl-3-[[(2RS)-2-(propylamino)propanoyl]amino]thiophene-2-carboxamide hydrochloride
![4-Methyl-N-propyl-3-[[(2RS)-2-(propylamino)propanoyl]amino]thiophene-2-carboxamide hydrochloride certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/277/493/7651cc5d-47a5-4017-836e-5a3c52875481/640/7651cc5d-47a5-4017-836e-5a3c52875481.png)
Synonym(s):
Articaine acid propionamide hydrochloride
Empirical Formula (Hill Notation):
C15H25N3O2S · HCl
Molecular Weight:
347.90
Products
Brand
Formula Weight
Manufacturer
Product Line
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1