4′-Aminobenzo-18-crown-6
Synonym(s):
(Benzo-18-crown-6)-4′-ylamine
Empirical Formula (Hill Notation):
C16H25NO6
CAS Number:
Molecular Weight:
327.37
Products
Brand
Formula Weight
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing