4,4′-(Hexafluoroisopropylidene)bis(p-phenyleneoxy)dianiline
Synonym(s):
2,2-Bis[4-(4-aminophenoxy)phenyl]hexafluoropropane, 4′,4′′′-(Hexafluoroisopropylidene)bis(4-phenoxyaniline)
Linear Formula:
(CF3)2C(C6H4OC6H4NH2)2
CAS Number:
Molecular Weight:
518.45
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing