![4,4-Diethoxy-N,N-dimethyl-1-butanamine](/deepweb/assets/sigmaaldrich/product/structures/107/279/ad608903-72bb-4b5a-a710-ec607eb10505/640/ad608903-72bb-4b5a-a710-ec607eb10505.png)
4,4-Diethoxy-N,N-dimethyl-1-butanamine
![4,4-Diethoxy-N,N-dimethyl-1-butanamine](/deepweb/assets/sigmaaldrich/product/structures/107/279/ad608903-72bb-4b5a-a710-ec607eb10505/640/ad608903-72bb-4b5a-a710-ec607eb10505.png)
Synonym(s):
1-(N,N-Dimethylamino)-4,4-diethoxybutane, 4,4-Diethoxy-N,N-dimethylbutanamine, 4,4-Diethoxy-N,N-dimethylbutylamine, 4-Dimethylaminobutanal diethyl acetal, N,N-Dimethyl-4-aminobutanal diethyl acetal
Empirical Formula (Hill Notation):
C10H23NO2
CAS Number:
Molecular Weight:
189.30
Products
Brand
Formula Weight
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing