Skip to Content
Merck
3-(Trifluoromethyl)phenylboronic acid ≥95%

3-(Trifluoromethyl)phenylboronic acid

3-(Trifluoromethyl)phenylboronic acid ≥95%
Synonym(s):
3-(Trifluoromethyl)benzeneboronic acid, a,a,a-Trifluoro-m-tolueneboronic acid
Linear Formula:
CF3C6H4B(OH)2
CAS Number:
Molecular Weight:
189.93

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing