Skip to Content
Merck
(3E)-4-Phenylbut-3-enoic acid methyl ester

(3E)-4-Phenylbut-3-enoic acid methyl ester

(3E)-4-Phenylbut-3-enoic acid methyl ester
Synonym(s):
Methyl (3E)-4-Phenylbut-3-enoate
Empirical Formula (Hill Notation):
C11H12O2
CAS Number:
Molecular Weight:
176.21

Products

Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing