Skip to Content
Merck
3,5-Dimethylbenzenethiol 90%

3,5-Dimethylbenzenethiol

3,5-Dimethylbenzenethiol 90%
Synonym(s):
3,5-Dimethylthiophenol
Linear Formula:
(CH3)2C6H3SH
CAS Number:
Molecular Weight:
138.23
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing