3,5-Bis(trifluoromethyl)benzaldehyde
Linear Formula:
(CF3)2C6H3CHO
CAS Number:
Molecular Weight:
242.12
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing