Skip to Content
Merck
3,3′,5,5′-Tetramethylbenzidine standard for GC

3,3′,5,5′-Tetramethylbenzidine

3,3′,5,5′-Tetramethylbenzidine standard for GC
Synonym(s):
BM blue, Sure Blue TMB, TMB, TMB Blotting Plus, TMB substrate, 3,3′,5,5′-Tetramethylbenzidine
Linear Formula:
[-C6H2(CH3)2-4-NH2]2
CAS Number:
Molecular Weight:
240.34
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique