Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
3,3′,5,5′-Tetramethylbenzidine
Products
Product Category
substrates (5)
bioactive small molecules (1)
Brand
Sigma-Aldrich (5)
Physical Form
Quality Segment
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
T0440
3,3′,5,5′-Tetramethylbenzidine,
peroxidase substrate, chromogenic, liquid
Expand
View Pricing
T8665
3,3′,5,5′-Tetramethylbenzidine,
peroxidase substrate, chromogenic, liquid
Expand
View Pricing
T0565
3,3′,5,5′-Tetramethylbenzidine,
TMB membrane substrate, chromogenic, liquid
Expand
View Pricing
T5569
3,3′,5,5′-Tetramethylbenzidine,
peroxidase substrate, chromogenic, liquid
Expand
View Pricing
T4319
3,3′,5,5′-Tetramethylbenzidine,
peroxidase substrate, chromogenic, liquid
Expand
View Pricing