Skip to Content
Merck
2-Methylbutyraldehyde analytical standard

2-Methylbutyraldehyde

2-Methylbutyraldehyde analytical standard
Linear Formula:
C2H5CH(CH3)CHO
CAS Number:
Molecular Weight:
86.13
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing