![2-Dodecanol 99%](/deepweb/assets/sigmaaldrich/product/structures/474/857/a9a9330e-483b-47e4-8aef-3cd35231bbf3/640/a9a9330e-483b-47e4-8aef-3cd35231bbf3.png)
2-Dodecanol
![2-Dodecanol 99%](/deepweb/assets/sigmaaldrich/product/structures/474/857/a9a9330e-483b-47e4-8aef-3cd35231bbf3/640/a9a9330e-483b-47e4-8aef-3cd35231bbf3.png)
Synonym(s):
Decyl methyl carbinol
Linear Formula:
CH3(CH2)9CH(OH)CH3
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing