![2-Chloro-2-deoxy-D-glucose analytical standard](/deepweb/assets/sigmaaldrich/product/structures/384/097/af3b9b29-dad3-4f43-97a5-d7e9536b0802/640/af3b9b29-dad3-4f43-97a5-d7e9536b0802.png)
2-Chloro-2-deoxy-D-glucose
![2-Chloro-2-deoxy-D-glucose analytical standard](/deepweb/assets/sigmaaldrich/product/structures/384/097/af3b9b29-dad3-4f43-97a5-d7e9536b0802/640/af3b9b29-dad3-4f43-97a5-d7e9536b0802.png)
Synonym(s):
2-CIDG
Empirical Formula (Hill Notation):
C6H11ClO5
CAS Number:
Molecular Weight:
198.60
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Format
Analyte Suitability
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing