![2-Butanone-2,4-DNPH certified reference material, vial of 100 mg, Cerilliant®](/deepweb/assets/sigmaaldrich/product/images/404/105/9dc355b8-8dc1-4eff-8de5-11f094b058cc/640/9dc355b8-8dc1-4eff-8de5-11f094b058cc.jpg)
2-Butanone-2,4-DNPH
![2-Butanone-2,4-DNPH certified reference material, vial of 100 mg, Cerilliant®](/deepweb/assets/sigmaaldrich/product/images/404/105/9dc355b8-8dc1-4eff-8de5-11f094b058cc/640/9dc355b8-8dc1-4eff-8de5-11f094b058cc.jpg)
Synonym(s):
2-Butanone-2,4-dinitrophenylhydrazone
Empirical Formula (Hill Notation):
C10H12N4O4
CAS Number:
Molecular Weight:
252.23
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing