Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
25-Hydroxyvitamin D
2
-(22,26,27-
13
C
3
) solution
Empirical Formula (Hill Notation):
13
C
3
C
25
H
44
O
2
Molecular Weight:
415.63
Products
Product Category
stable isotopes (2)
vitamins (2)
Brand
Sigma-Aldrich (2)
Formula Weight
Purity
Quality Segment
Isotope
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
924903
25-Hydroxyvitamin D
2
-(22,26,27-
13
C
3
) solution,
50 μg/mL in ethanol, ≥98 atom %
13
C, ≥95% (CP)
Expand
View Pricing
924881
25-Hydroxyvitamin D
2
-(22,26,27-
13
C
3
) solution,
10 μg/mL in ethanol, ≥98 atom %
13
C, ≥95% (CP)
Expand
View Pricing