Skip to Content
Merck
2,4-Dimethylaniline certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

2,4-Dimethylaniline

2,4-Dimethylaniline certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
1-Amino-2,4-dimethylbenzene, 2,4-Xylidine, 4-Amino-m-xylene
Linear Formula:
(CH3)2C6H3NH2
CAS Number:
Molecular Weight:
121.18

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Markush Class
Markush Group