
2,4,6-Tri-tert-butylpyridine

Empirical Formula (Hill Notation):
C17H29N
CAS Number:
Molecular Weight:
247.42
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1