Skip to Content
Merck
2,3,5,6-Tetramethylpyrazine analytical standard

2,3,5,6-Tetramethylpyrazine

2,3,5,6-Tetramethylpyrazine analytical standard
Empirical Formula (Hill Notation):
C8H12N2
CAS Number:
Molecular Weight:
136.19
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique