![2,3,4-Trichlorophenol PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/241/345/714599ec-c6f9-494c-bd93-17874a912db8/640/714599ec-c6f9-494c-bd93-17874a912db8.png)
2,3,4-Trichlorophenol
![2,3,4-Trichlorophenol PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/241/345/714599ec-c6f9-494c-bd93-17874a912db8/640/714599ec-c6f9-494c-bd93-17874a912db8.png)
Linear Formula:
Cl3C6H2OH
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing