2,3,4,5-Tetrafluorobenzoic acid
Linear Formula:
HC6F4CO2H
CAS Number:
Molecular Weight:
194.08
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing